2-Chloro-4,5-diamino-6-methylpyrimidine 97%

Supplier: Ambeed
Warning

A116892-1G A116892-250MG A116892-5G
77257-970EA 35 USD
77257-970 77257-972 77257-974
2-Chloro-4,5-diamino-6-methylpyrimidine 97%
2-Chloro-4,5-diamino-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₇ClN₄
MDL Number: MFCD01764685
CAS Number: 63211-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR