6-Iodouracil 95%

Supplier: Ambeed
Warning

A116789-250MG A116789-5G
77284-564EA 11 USD
77284-564 77747-849
6-Iodouracil 95%
6-Iodouracil

Precaución: for Research Use Only

Formula: C₄H₃IN₂O₂
MW: 237.9800 g/mol
MDL Number: MFCD01346695
CAS Number: 4269-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR