3-Chloro-5-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A116722-10G A116722-1G A116722-250MG A116722-5G A116722-25G
77271-238EA 53 USD
77271-238 77271-240 77271-242 77271-244 77566-122
3-Chloro-5-iodobenzoic acid 98%
3-Chloro-5-iodobenzoic acid

Precaución: For research use only.

Formula: C₇H₄ClIO₂
MDL Number: MFCD00672966
CAS Number: 289039-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR