4,8-Bis[5-(2-ethylhexyl)thiophen-2-yl]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene 98%

Supplier: Ambeed
Danger

A116715-100MG A116715-1G A116715-250MG A116715-5G
77257-956EA 23 USD
77257-956 77257-958 77257-960 77257-962
4,8-Bis[5-(2-ethylhexyl)thiophen-2-yl]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene 98%
4,8-Bis[5-(2-ethylhexyl)thiophen-2-yl]-2,6-bis(trimethylstannyl)benzo[1,2-b:4,5-b']dithiophene

Precaución: for Research Use Only

Formula: C₄₀H₅₈S₄Sn₂
MDL Number: MFCD28386086
CAS Number: 1352642-37-5
UN: 3146
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR