3-Chloromethyl-5-methyl[1,2,4]oxadiazole 98%

Supplier: Ambeed
Danger

A116661-10G A116661-1G A116661-250MG
77271-228EA 245 USD
77271-228 77271-230 77271-232
3-Chloromethyl-5-methyl[1,2,4]oxadiazole 98%
3-Chloromethyl-5-methyl[1,2,4]oxadiazole

Precaución: for Research Use Only

Formula: C₄H₅ClN₂O
MDL Number: MFCD01809083
CAS Number: 1192-80-9
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR