3-(Dicyanomethylidene)indan-1-one 97%

Supplier: Ambeed
Danger

Synonyms: 2-(3-Oxo-2,3-dihydro-1H-inden-1-ylidene)malononitrile

A116566-100MG A116566-1G A116566-250MG A116566-5G
77271-220EA 14 USD
77271-220 77271-222 77271-224 77747-848
3-(Dicyanomethylidene)indan-1-one 97%
3-(Dicyanomethylidene)indan-1-one

Precaución: for Research Use Only

Formula: C₁₂H₆N₂O
MW: 194.19 g/mol
MDL Number: MFCD00143117
CAS Number: 1080-74-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR