4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: BPADA, 4,4'-(4,4'-Isopropylidenediphenoxy)bis(phthalic anhydride), 5,5'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(isobenzofuran-1,3-dione)

A116386-1000G A116386-100G A116386-10G A116386-25G A116386-500G A116386-5G
77284-544EA 478.95 USD
77284-544 77284-546 77284-548 77284-550 77284-552 77284-554
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride 97%
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride

Precaución: for Research Use Only

Formula: C₃₁H₂₀O₈
MW: 520.49 g/mol
MDL Number: MFCD00319167
CAS Number: 38103-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR