4-Amino-3-bromo-5-iodopyridine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-iodopyridin-4-amine, 3-Bromo-5-iodo-4-pyridylamine

A116340-1G A116340-250MG A116340-10G A116340-5G
77271-212EA 24.72 USD
77271-212 77271-214 77570-994 77747-846
4-Amino-3-bromo-5-iodopyridine 95%
4-Amino-3-bromo-5-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MW: 298.91 g/mol
MDL Number: MFCD07781147
CAS Number: 902837-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR