Di(naphthalen-1-yl)phosphine oxide 97%

Supplier: Ambeed
Warning

A116260-1G A116260-250MG A116260-5G A116260-25G
77233-930EA 28 USD
77233-930 77233-932 77233-934 77566-102
Di(naphthalen-1-yl)phosphine oxide 97%
Di(naphthalen-1-yl)phosphine oxide

Precaución: For research use only.

Formula: C₂₀H₁₅OP
MDL Number: MFCD30342487
CAS Number: 13440-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR