(S)-tert-Butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

A116133-1G A116133-250MG A116133-5G
77271-198EA 49.44 USD
77271-198 77271-200 77271-202
(S)-tert-Butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 97%
(S)-tert-Butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₀ClNO₂
MW: 269.76 g/mol
MDL Number: MFCD00672723
CAS Number: 82586-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR