4,4'-Dimethylbiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Bitolyl, 4,4'-Dimethyldiphenyl

A116118-1000G A116118-100G A116118-10G A116118-25G A116118-500G
77284-528EA 113.3 USD
77284-528 77284-530 77284-532 77284-534 77284-536
4,4'-Dimethylbiphenyl 98%
4,4'-Dimethylbiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄
MW: 182.27 g/mol
MDL Number: MFCD00008545
CAS Number: 613-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR