tert-Butyl 4-((2-methoxyethyl)amino)piperidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A116008-100MG
A116008-1G
A116008-250MG
A116008-25G
77284-520EA
15
USD
77284-520
77284-522
77284-524
77284-526
tert-Butyl 4-((2-methoxyethyl)amino)piperidine-1-carboxylate 97%
tert-Butyl 4-((2-methoxyethyl)amino)piperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₂₆N₂O₃ |
MDL Number:
MFCD06656630 CAS Number: 710972-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...