2-Butylimidazole 97%

Supplier: Ambeed
Warning

A115981-100G A115981-25G A115981-5G
77257-946EA 244.11 USD
77257-946 77257-948 77257-950
2-Butylimidazole 97%
2-Butylimidazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MW: 124.1800 g/mol
MDL Number: MFCD00792664
CAS Number: 50790-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR