2-Bromo-3-methoxyaniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-m-anisidine, 3-Amino-2-bromoanisole

A115888-10G A115888-1G A115888-250MG A115888-25G A115888-5G
77257-936EA 133.9 USD
77257-936 77257-938 77257-940 77257-942 77257-944
2-Bromo-3-methoxyaniline 98%
2-Bromo-3-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
MDL Number: MFCD07787450
CAS Number: 112970-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR