1-Butyl-3-methylpyridiniumhexafluorophosphate 99%
Supplier: Ambeed
|
Warning
|
A115869-25G
A115869-5G
77257-930EA
21
USD
77257-930
77257-932
1-Butyl-3-methylpyridiniumhexafluorophosphate 99%
1-Butyl-3-methylpyridiniumhexafluorophosphate
Precaución: for Research Use Only
Formula:
C₁₀H₁₆F₆NP |
MDL Number:
MFCD08458504 CAS Number: 845835-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...