3-Fluoro-4-methoxyphenol 98%

Supplier: Ambeed
Warning

A115794-1G A115794-250MG A115794-25G A115794-10G A115794-5G
77271-192EA 12 USD
77271-192 77271-194 77570-960 77614-906 77614-908
3-Fluoro-4-methoxyphenol 98%
3-Fluoro-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MDL Number: MFCD09263471
CAS Number: 452-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR