6-Bromo-3-methoxysalicylaldehyde 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-2-hydroxy-3-methoxybenzaldehyde
A115787-1G
A115787-250MG
A115787-25G
A115787-50MG
A115787-5G
A115787-10G
A115787-10MG
77233-914EA
17.51
USD
77233-914
77233-916
77233-918
77233-920
77233-922
77614-904
77745-856
6-Bromo-3-methoxysalicylaldehyde 97%
6-Bromo-3-methoxysalicylaldehyde
Precaución: for Research Use Only
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol |
MDL Number:
MFCD00046152 CAS Number: 20035-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...