Methyl 3-(3-(tert-butyl)-4-hydroxyphenyl)propanoate 97%

Supplier: Ambeed
Warning

A115768-10G A115768-5G
77245-904EA 27.81 USD
77245-904 77245-906
Methyl 3-(3-(tert-butyl)-4-hydroxyphenyl)propanoate 97%
Methyl 3-(3-(tert-butyl)-4-hydroxyphenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₃
MW: 236.31 g/mol
MDL Number: MFCD19441367
CAS Number: 36837-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR