4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Difluoro(3,4,5-trifluorophenoxy)methyl)-4'-ethyl-3,5-difluoro-1,1'-biphenyl

A115754-1G A115754-250MG A115754-25G A115754-5G
77233-906EA 14.42 USD
77233-906 77233-908 77233-910 77233-912
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl 97%
4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl

Precaución: for Research Use Only

Formula: C₂₁H₁₃F₇O
MW: 414.32 g/mol
MDL Number: MFCD22376741
CAS Number: 303186-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR