5-Bromouridine 98%

Supplier: Ambeed
Warning

A115724-10G A115724-1G A115724-250MG A115724-25G A115724-5G
77245-896EA 174.07 USD
77245-896 77245-898 77245-900 77245-902 77745-854
5-Bromouridine 98%
5-Bromouridine

Precaución: for Research Use Only

Formula: C₉H₁₁BrN₂O₆
MW: 323.10 g/mol
MDL Number: MFCD00006528
CAS Number: 957-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR