(3-Bromopropyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-phenylpropane

A115699-100G A115699-10G A115699-250MG A115699-25G A115699-500G A115699-5G
77233-892EA 45 USD
77233-892 77233-894 77233-896 77233-898 77233-900 77233-902 77233-904
(3-Bromopropyl)benzene 95%
(3-Bromopropyl)benzene

Precaución: for Research Use Only

Formula: BrH₂CCH₂CH₂(C₆H₅)
MDL Number: MFCD00000257
CAS Number: 637-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR