Ethyl-2-amino-5-bromobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-5-bromobenzoate

A115633-10G A115633-1G A115633-250MG A115633-25G
77245-890EA 131.84 USD
77245-890 77245-892 77245-894 77745-853
Ethyl-2-amino-5-bromobenzoate 98%
Ethyl-2-amino-5-bromobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.0900 g/mol
MDL Number: MFCD03428545
CAS Number: 63243-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR