6-Fluoropyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoropicolinonitrile, 2-Cyano-6-fluoropyridine

A115488-10G A115488-1G A115488-250MG A115488-25G A115488-5G
77257-916EA 101.97 USD
77257-916 77257-918 77257-920 77257-922 77257-924
6-Fluoropyridine-2-carbonitrile 97%
6-Fluoropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD09263323
CAS Number: 3939-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR