4-Hydroxycoumarin 98%

Supplier: Ambeed
Warning

A115444-1000G A115444-100G A115444-10G A115444-25G A115444-500G
77257-900EA 105.7 USD
77257-900 77257-902 77257-904 77257-906 77257-908
4-Hydroxycoumarin 98%
4-Hydroxycoumarin

Precaución: for Research Use Only

Formula: C₉H₆O₃
MDL Number: MFCD00006856
CAS Number: 1076-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR