5-Bromo-2-fluoroaniline 98%

Supplier: Ambeed
Warning

A115330-100G A115330-10G A115330-1G A115330-25G A115330-500G A115330-5G
77284-472EA 53.56 USD
77284-472 77284-474 77284-476 77284-478 77284-480 77284-482
5-Bromo-2-fluoroaniline 98%
5-Bromo-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03094181
CAS Number: 2924-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR