5-Chloro-2,3-dihydro-1H-inden-1-amine 95%

Supplier: Ambeed
Warning

A115190-5G A115190-250MG
77233-864EA 303 USD
77233-864 77745-851
5-Chloro-2,3-dihydro-1H-inden-1-amine 95%
5-Chloro-2,3-dihydro-1H-inden-1-amine

Precaución: for Research Use Only

Formula: C₉H₁₀ClN
MDL Number: MFCD07373935
CAS Number: 67120-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR