LY-2090314 99%

Supplier: Ambeed
Warning

Synonyms: 3-(9-fluoro-2-(piperidine-1-carbonyl)-1,2,3,4-tetrahydro-[1,4]diazepino[6,7,1-hi]indol-7-yl)-4-(imidazo[1,2-a]pyridin-3-yl)-1H-pyrrole-2,5-dione, LY2090314

A115111-100MG A115111-1MG A115111-50MG A115111-5MG
77233-856EA 233.81 USD
77233-856 77233-858 77233-860 77233-862
LY-2090314 99%
LY-2090314

Precaución: for Research Use Only

Formula: C₂₈H₂₅FN₆O₃
MW: 512.53 g/mol
MDL Number: MFCD20526532
CAS Number: 603288-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR