4-Bromo-2,6-dichlorophenol 98%

Supplier: Ambeed
Warning

A115050-100G A115050-10G A115050-1G A115050-25G A115050-5G
77284-450EA 55 USD
77284-450 77284-452 77284-454 77284-456 77284-458
4-Bromo-2,6-dichlorophenol 98%
4-Bromo-2,6-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MDL Number: MFCD00019996
CAS Number: 3217-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR