(2-Chlorophenyl)acetone 97%

Supplier: Ambeed
Warning

A114978-10G A114978-1G
77257-868EA 51.01 USD
77257-868 77257-870
(2-Chlorophenyl)acetone 97%
(2-Chlorophenyl)acetone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MDL Number: MFCD00045175
CAS Number: 6305-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR