Amodiaquine dihydrochloride 98%

Supplier: Ambeed
Warning

A114970-1000G A114970-100G A114970-10G A114970-25G A114970-25MG A114970-500G A114970-5G A114970-5MG
77284-424EA 348.99 USD
77284-424 77284-426 77284-428 77284-430 77284-432 77284-434 77284-436 77284-438
Amodiaquine dihydrochloride 98%
Amodiaquine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₄Cl₃N₃O
MDL Number: MFCD00078857
CAS Number: 69-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR