(4-(2-Hydroxyethoxy)phenyl)(phenyl)methanone 97%

Supplier: Ambeed
Warning

A114894-100MG A114894-1G A114894-250MG
77245-866EA 5 USD
77245-866 77245-868 77245-870
(4-(2-Hydroxyethoxy)phenyl)(phenyl)methanone 97%
(4-(2-Hydroxyethoxy)phenyl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00623913
CAS Number: 14814-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR