2-Oxo-2,3-dihydro-1H-benzo[d]imidazole-5-carbaldehyde 98%

Supplier: Ambeed
Warning

A114826-100MG A114826-1G A114826-250MG
77233-840EA 78.28 USD
77233-840 77233-842 77233-844
2-Oxo-2,3-dihydro-1H-benzo[d]imidazole-5-carbaldehyde 98%
2-Oxo-2,3-dihydro-1H-benzo[d]imidazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MW: 162.14 g/mol
MDL Number: MFCD18807917
CAS Number: 106429-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR