1,2,4-Triazole-3-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: 2H-1,2,4-Triazole-3-carboxamide

A114764-1G A114764-25G A114764-5G
77233-834EA 8.24 USD
77233-834 77233-836 77233-838
1,2,4-Triazole-3-carboxamide 98%
1,2,4-Triazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₃H₄N₄O
MW: 112.09 g/mol
MDL Number: MFCD03990481
CAS Number: 3641-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR