2-Ethyl-3-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A114705-10G A114705-1G A114705-250MG
77271-152EA 115.36 USD
77271-152 77271-154 77271-156
2-Ethyl-3-methoxybenzoic acid 98%
2-Ethyl-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MDL Number: MFCD11100772
CAS Number: 57598-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR