2-(3-Bromopropoxy)tetrahydropyran 94% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(3-Bromopropoxy)tetrahydro-2H-pyran
Stabilization:
Stabilised with potassium carbonate 0.5%
A114702-1G
A114702-25G
A114702-10G
A114702-5G
77233-830EA
6
USD
77233-830
77233-832
77549-256
77745-849
2-(3-Bromopropoxy)tetrahydropyran 94% stabilised
2-(3-Bromopropoxy)tetrahydropyran
Precaución: for Research Use Only
Formula:
C₈H₁₅BrO₂ |
MDL Number:
MFCD00058593 CAS Number: 33821-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...