4-Bromo-3-methylbenzamide 98%

Supplier: Ambeed
Warning

A114674-100G A114674-1G A114674-25G
77233-824EA 185 USD
77233-824 77233-826 77233-828
4-Bromo-3-methylbenzamide 98%
4-Bromo-3-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD00672914
CAS Number: 170229-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR