Adenine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 7H-Purin-6-amine hydrochloride, Purin-6-amine hydrochloride, 6-Aminopurine hydrochloride

A114669-100G A114669-100MG A114669-25G A114669-500G
77271-136EA 61.8 USD
77271-136 77271-138 77271-140 77271-142
Adenine hydrochloride 98%
Adenine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₅N₅·HCl
MW: 171.59 g/mol
MDL Number: MFCD00150864
CAS Number: 2922-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR