3-Fluoro-5-iodoaniline 97%

Supplier: Ambeed
Warning

A114613-10G A114613-1G A114613-5G
77271-130EA 160 USD
77271-130 77271-132 77271-134
3-Fluoro-5-iodoaniline 97%
3-Fluoro-5-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅FIN
MDL Number: MFCD04116466
CAS Number: 660-49-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR