3-(2-Bromoacetyl)-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one 97%

Supplier: Ambeed
Danger

Synonyms: 3-(2-Bromoacetyl)-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one

A114554-100MG A114554-10G A114554-250MG A114554-25G A114554-5G
77233-808EA 17.51 USD
77233-808 77233-810 77233-812 77233-814 77233-816
3-(2-Bromoacetyl)-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one 97%
3-(2-Bromoacetyl)-10,11-dihydro-5H-dibenzo[c,g]chromen-8(9H)-one

Precaución: for Research Use Only

Formula: C₁₉H₁₅BrO₃
MW: 371.22 g/mol
MDL Number: MFCD30187855
CAS Number: 1378390-29-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR