4-(3-Cyanophenyl)morpholine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Morpholinobenzonitrile, 3-(Morpholin-4-yl)benzonitrile

A114539-1G A114539-250MG
77284-404EA 19.57 USD
77284-404 77284-406
4-(3-Cyanophenyl)morpholine 97%
4-(3-Cyanophenyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O
MW: 188.2300 g/mol
MDL Number: MFCD07368523
CAS Number: 204078-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR