N-(Pyridin-4-yl)pyridine-4-carboxamide 97%

Supplier: Ambeed
Warning

Synonyms: N-(Pyridin-4-yl)isonicotinamide

A114514-100MG A114514-1G A114514-5G A114514-250MG
77245-856EA 40.17 USD
77245-856 77245-858 77245-860 77745-847
N-(Pyridin-4-yl)pyridine-4-carboxamide 97%
N-(Pyridin-4-yl)pyridine-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₁H₉N₃O
MW: 199.209 g/mol
MDL Number: MFCD00454168
CAS Number: 64479-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR