(3,5-Difluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A114463-10G A114463-1G A114463-250MG A114463-100G A114463-25G
77233-802EA 19 USD
77233-802 77233-804 77233-806 77566-014 77570-814
(3,5-Difluorophenyl)acetic acid 98%
(3,5-Difluorophenyl)acetic acid

Precaución: For research use only.

Formula: (C₆H₃F₂)CH₂COOH
MDL Number: MFCD00010316
CAS Number: 105184-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR