2-Amino-N,N-dimethylacetamide hydrochloride 97%

Supplier: Ambeed
Warning

A114456-10G A114456-1G A114456-250MG A114456-25G A114456-5G
77257-824EA 70 USD
77257-824 77257-826 77257-828 77257-830 77257-832
2-Amino-N,N-dimethylacetamide hydrochloride 97%
2-Amino-N,N-dimethylacetamide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂O
MDL Number: MFCD08060117
CAS Number: 72287-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR