L(+)-Ribose 97%

Supplier: Ambeed
Warning

A114424-10G A114424-1G A114424-25G A114424-5G A114424-100G A114424-500G
77284-382EA 13 USD
77284-382 77284-384 77284-386 77284-388 77592-220 77614-894
L(+)-Ribose 97%
L(+)-Ribose

Precaución: for Research Use Only

Formula: C₅H₁₀O₅
MDL Number: MFCD00167010
CAS Number: 24259-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR