3,4-Dichloropyridin-2-amine 96%

Supplier: Ambeed
Warning

A114329-100MG A114329-1G A114329-250MG A114329-5G
77245-848EA 13 USD
77245-848 77245-850 77245-852 77245-854
3,4-Dichloropyridin-2-amine 96%
3,4-Dichloropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MDL Number: MFCD08272175
CAS Number: 188577-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR