4-Bromo-1-butanol 80-85% (contains varying amounts of Tetrahydrofuran)

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-4-hydroxybutane

A114318-10G A114318-1G A114318-25G A114318-5G
77284-366EA 31 USD
77284-366 77284-368 77284-370 77284-372
4-Bromo-1-butanol 80-85% (contains varying amounts of Tetrahydrofuran)
4-Bromo-1-butanol

Precaución: for Research Use Only

Formula: BrH₂C(CH₂)₂CH₂OH
MW: 153.02
MDL Number: MFCD00015387
CAS Number: 33036-62-3
UN: 1987
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR