4-Bromo-2,3-difluoroaniline 97%
Supplier: Ambeed
|
Warning
|
A114207-100MG
A114207-10G
A114207-1G
A114207-250MG
A114207-25G
A114207-100G
A114207-5G
77284-356EA
6.18
USD
77284-356
77284-358
77284-360
77284-362
77284-364
77747-828
77747-829
4-Bromo-2,3-difluoroaniline 97%
4-Bromo-2,3-difluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrF₂N MW: 208.0035 g/mol |
MDL Number:
MFCD09030643 CAS Number: 112279-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...