4-Amino-4H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1,2,4-triazole, 1,2,4-Triazol-4-ylamine, 4H-1,2,4-Triazol-4-amine

A114038-100G A114038-10G A114038-1G A114038-25G A114038-500G A114038-5G
77284-322EA 18 USD
77284-322 77284-324 77284-326 77284-328 77284-330 77284-332
4-Amino-4H-1,2,4-triazole 98%
4-Amino-4H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₂H₄N₄
MW: 84.08 g/mol
MDL Number: MFCD00003099
CAS Number: 584-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR