Lifitegrast 97%

Supplier: Ambeed
Warning

Synonyms: SAR-1118, Lifitegrast, Xiidra., (S)-2-(2-(benzofuran-6-carbonyl)-5,7-dichloro-1,2,3,4-tetrahydroisoquinoline-6-carboxamido)-3-(3-(methylsulphonyl)phenyl)propanoic acid, SAR 1118, SAR1118

A113895-10MG A113895-1MG A113895-25MG A113895-50MG A113895-5MG A113895-1G
77257-808EA 11.33 USD
77257-808 77257-810 77257-812 77257-814 77257-816 77570-756
Lifitegrast 97%
Lifitegrast

Precaución: for Research Use Only

Formula: C₂₉H₂₄Cl₂N₂O₇S
MW: 615.48 g/mol
MDL Number: MFCD28502439
CAS Number: 1025967-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR