1,3-Difluoro-5-(trans-4-propylcyclohexyl)benzene 97%

Supplier: Ambeed
Warning

A113781-10G A113781-1G A113781-5G
77233-782EA 27.81 USD
77233-782 77233-784 77233-786
1,3-Difluoro-5-(trans-4-propylcyclohexyl)benzene 97%
1,3-Difluoro-5-(trans-4-propylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₅H₂₀F₂
MW: 238.32 g/mol
MDL Number: MFCD09750933
CAS Number: 144261-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR